missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylpiperazine Dihydrochloride, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 1-Methylpiperazine Dihydrochloride |
| CAS | 34352-59-5 |
| Molecular Formula | C5H14Cl2N2 |
| Quantity | 10 g |
| Synonym | N-Methylpiperazine Dihydrochloride |
| InChI Formula | InChI=1S/C5H12N2.2ClH/c1-7-4-2-6-3-5-7;;/h6H,2-5H2,1H3;2*1H |
| SMILES | Cl.Cl.CN1CCNCC1 |
| IUPAC Name | 1-methylpiperazine;dihydrochloride |
| Molecular Weight (g/mol) | 173.08 |
| Formula Weight | 172.0534 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction