missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-3-methylimidazolium n-octyl sulfate, 99%

Product Code. 11312849
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312849 50 g 50g
11302849 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312849 Supplier Thermo Scientific Alfa Aesar Supplier No. H27720.18
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 445473-58-5 | C16H32N2O4S | 348.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 445473-58-5
Molecular Formula C16H32N2O4S
Molecular Weight (g/mol) 348.50
MDL Number MFCD03788914
InChI Key KIDIBVPFLKLKAH-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium octyl sulfate, 1-butyl-3-methylimidazolium octylsulfate, 1-butyl-3-methyl-1h-imidazol-3-ium octyl sulfate, 1-butyl-3-methylimidazolium n-octyl sulfate, acmc-20aowg, bmim osu, 1-methyl-3-butylimidazolium octyl sulfate, 1-n-butyl-3-methylimidazoliumoctyl sulfate, 1-butyl-3-methylimidazolium 1-octyl sulfate, 1-n-butyl-3-methylimidazolium octyl sulfate
PubChem CID 12095226
IUPAC Name 1-butyl-3-methylimidazol-3-ium;octyl sulfate
SMILES CCCCN1C=C[N+](C)=C1.CCCCCCCCOS([O-])(=O)=O

Specifications

Quantity 50 g
Formula Weight 348.5
Percent Purity 99%
Chemical Name or Material 1-n-Butyl-3-methylimidazolium n-octyl sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.