missing translation for 'onlineSavingsMsg'
Learn More

1-Nonanol, TRC

Product Code. 30480707
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480707 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30480707 Supplier Toronto Research Chemicals Supplier No. TRCN64934050ML
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Nonanol
CAS 143-08-8
Molecular Formula C9 H20 O
Quantity 50 mL
Synonym 1-Nonanol (9CI','ACI), Nonyl alcohol (8CI), 1-Hydroxynonane, Linevol 9, n-Nonan-1-ol, n-Nonyl alcohol, Nonan-1-ol, Nonanol, NSC 5521, Octyl carbinol, Pelargonic alcohol
InChI Formula InChI=1S/C9H20O/c1-2-3-4-5-6-7-8-9-10/h10H,2-9H2,1H3
SMILES CCCCCCCCCO
IUPAC Name nonan-1-ol
Molecular Weight (g/mol) 144.25
Formula Weight 144.1514
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.