missing translation for 'onlineSavingsMsg'
Learn More

1-O-Dimethoxytrityl-2-(N-Fmoc)-4-Aminobutyl)-1,3-propanediol, >95%, TRC

Product Code. 17998224
Change view
Click to view available options
Quantity:
100 mg
250 mg
2.5 g
Unit Size:
100mg
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17998224 100 mg 100mg
17937734 2.5 g 2.5g
17937644 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17998224 Supplier Toronto Research Chemicals Supplier No. TRCD460760100MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147190-32-7
Molecular Formula C43 H45 N O6
Molecular Weight (g/mol) 671.82
Synonym [6-[Bis(4-methoxyphenyl)phenylmethoxy]-5-(hydroxymethyl)hexyl]carbamic Acid 9H-fluoren-9-ylmethyl Ester, Diethyl Ester (4-Chlorobutyl)propanedioic Acid, Diethyl Ester (4-Chlorobutyl)malonic Acid, 1,3-Diethyl 2-(4-Chlorobutyl)propanedioate
IUPAC Name 9H-fluoren-9-ylmethyl N-[5-[[bis(4-methoxyphenyl)-phenyl-methoxy]methyl]-6-hydroxy-hexyl]carbamate
SMILES OCC(CCCCNC(OCC1C2=C(C=CC=C2)C3=C1C=CC=C3)=O)COC(C4=CC=CC=C4)(C5=CC=C(OC)C=C5)C6=CC=C(OC)C=C6

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C43H45NO6/c1-47-35-23-19-33(20-24-35)43(32-13-4-3-5-14-32,34-21-25-36(48-2)26-22-34)50-29-31(28-45)12-10-11-27-44-42(46)49-30-41-39-17-8-6-15-37(39)38-16-7-9-18-40(38)41/h3-9,13-26,31,41,45H,10-12,27-30H2,1-2H3,(H,44,46)
Formula Weight 671.3247
Percent Purity >95%
Chemical Name or Material 1-O-Dimethoxytrityl-2-(N-Fmoc)-4-Aminobutyl-1,3-propanediol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.