missing translation for 'onlineSavingsMsg'
Learn More

1-O-Hexyl-2,3,5-trimethylhydroquinone, TRC

Product Code. 30396090
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396090 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396090 Supplier Toronto Research Chemicals Supplier No. TRCH325000100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 148081-72-5
Molecular Formula C15 H26 O2
Quantity 100 mg
InChI Formula InChI=1S/C15H26O2/c1-5-6-7-8-9-17-14-10-11(2)15(16)13(4)12(14)3/h10,14-16H,5-9H2,1-4H3
SMILES CCCCCCOC1C=C(C)C(O)C(=C1C)C
IUPAC Name 4-hexoxy-2,3,6-trimethylcyclohexa-2,5-dien-1-ol
Molecular Weight (g/mol) 238.37
Formula Weight 238.1933
Chemical Name or Material 1-O-Hexyl-2,3,5-trimethylhydroquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.