Learn More
(1-Octyl)triphenylphosphonium bromide, 97%
Product Code. 11397695
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397695 100 g 100g
11377695 5 g 5g
11387695 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11397695 Supplier Thermo Scientific Alfa Aesar Supplier No. L02412.22
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 42036-78-2 | C26H32BrP | 455.42 g/mol
(1-Octyl)triphenylphosphonium bromide is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(1-Octyl)triphenylphosphonium bromide is used as a pharmaceutical intermediate.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 42036-78-2
Molecular Formula C26H32BrP
Molecular Weight (g/mol) 455.42
MDL Number MFCD00051874
InChI Key OBLXVLWZBMAMHE-UHFFFAOYSA-M
Synonym octyltriphenylphosphonium bromide, 1-octyl triphenylphosphonium bromide, n-octyl triphenylphosphonium bromide, n-octyltriphenylphosphonium bromide, octyltriphenylphosphanium bromide, acmc-1ao06, octyl triphenyl phosphanium bromide, n-octyl-triphenylphosphonium bromide, phosphonium,octyltriphenyl-, bromide 1:1, 1-octyl triphenyl phosphonium bromide
PubChem CID 3084855
IUPAC Name octyl(triphenyl)phosphanium;bromide
SMILES [Br-].CCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Specifications
Melting Point 61°C to 63°C
Linear Formula CH3(CH2)7P(C6H5)3Br
Quantity 100 g
Beilstein 3642460
Sensitivity Hygroscopic
Formula Weight 455.42
Percent Purity 97%
Chemical Name or Material (1-Octyl)triphenylphosphonium bromide