missing translation for 'onlineSavingsMsg'
Learn More

1-Pentadecanethiol, TRC

Product Code. 30410318
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30410318 100 mg
30418060 500 mg
30409167 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30410318 Supplier Toronto Research Chemicals Supplier No. TRCP267983100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25276-70-4
Molecular Formula C15H32S
Molecular Weight (g/mol) 244.48
IUPAC Name pentadecane-1-thiol
SMILES CCCCCCCCCCCCCCCS

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H32S/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16/h16H,2-15H2,1H3
Formula Weight 244.2225
Chemical Name or Material 1-Pentadecanethiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.