missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanesulfonic acid, sodium salt hydrate, 99%

Product Code. 10318953 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10318953 100 g Glass bottle 100g
10169322 5 g Glass bottle 5g
10023230 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10318953 Supplier Thermo Scientific Acros Supplier No. 270371000
  • ONLINE EXCLUSIVE PRICE
    525.00€
    508.00€ / 100g
    Save 17.00€
    3% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1266615-65-9 | C5H11NaO3S | 174.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1266615-65-9
Molecular Formula C5H11NaO3S
Molecular Weight (g/mol) 174.19
MDL Number MFCD00149548
InChI Key ROBLTDOHDSGGDT-UHFFFAOYSA-M
Synonym n-Amylsulfonic acid, sodium salt
IUPAC Name sodium pentane-1-sulfonate
SMILES [Na+].CCCCCS([O-])(=O)=O

Specifications

Melting Point >300.0°C
Color White
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99%
Linear Formula CH3(CH2)4SO3Na·xH2O
Beilstein 04, III, 23
Solubility Information (10% in water) Clear colorless
Formula Weight 174.12
Percent Purity 99%
Infrared Spectrum Authentic
Loss on Drying 9.6% max. (100°C, 3 hrs) (vacuum dried)
Packaging Glass bottle
Appearance (10% aq. soln.) clear, colorless
Chemical Name or Material 1-Pentanesulfonic acid, sodium salt hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.