missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylcyclopentanecarboxylic Acid, Mikromol™

Product Code. 30051600
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30051600

Brand: Mikromol MM1268.010025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-Phenylcyclopentanecarboxylic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Phenylcyclopentanecarboxylic Acid
CAS 77-55-4
Analyte or Component Names 1-Phenylcyclopentanecarboxylic Acid
Shipping Condition Room Temperature
Molecular Formula C12 H14 O2
Certifications/Compliance ISO 9001
Synonym Pentoxyverine Hydrogen Citrate Imp. A (EP), Pentoxyverine Imp. A (EP), 1-Phenylcyclopentanecarboxylic Acid, Pentoxyverine Hydrogen Citrate Impurity A, Pentoxyverine Impurity A
InChI Formula InChI=1S/C12H14O2/c13-11(14)12(8-4-5-9-12)10-6-2-1-3-7-10/h1-3,6-7H,4-5,8-9H2,(H,13,14)
SMILES OC(=O)C1(CCCC1)c2ccccc2
IUPAC Name 1-phenylcyclopentane-1-carboxylic acid
Molecular Weight (g/mol) 190.24
Quantity 25 mg
Formula Weight 190.0994 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.