missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylcyclopentanecarboxylic Acid, Mikromol™

Product Code. 30052249
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052249 100 mg 100mg
30051600 25 mg 25mg
30460392 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30052249 Supplier Mikromol Supplier No. MM1268.01
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-Phenylcyclopentanecarboxylic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Phenylcyclopentanecarboxylic Acid
CAS 77-55-4
Analyte or Component Names 1-Phenylcyclopentanecarboxylic Acid
Shipping Condition Room Temperature
Molecular Formula C12 H14 O2
Certifications/Compliance ISO 9001
Synonym Pentoxyverine Hydrogen Citrate Imp. A (EP), Pentoxyverine Imp. A (EP), 1-Phenylcyclopentanecarboxylic Acid, Pentoxyverine Hydrogen Citrate Impurity A, Pentoxyverine Impurity A
InChI Formula InChI=1S/C12H14O2/c13-11(14)12(8-4-5-9-12)10-6-2-1-3-7-10/h1-3,6-7H,4-5,8-9H2,(H,13,14)
SMILES OC(=O)C1(CCCC1)c2ccccc2
IUPAC Name 1-phenylcyclopentane-1-carboxylic acid
Molecular Weight (g/mol) 190.24
Quantity 100 mg
Formula Weight 190.0994 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.