missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylcyclopropanecarboxylic acid, 97%

Product Code. 11411800
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11411800 5 g 5g
11421800 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11411800 Supplier Thermo Scientific Alfa Aesar Supplier No. H27525.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6120-95-2 | C10H10O2 | 162.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6120-95-2
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00001288
InChI Key IWWCCNVRNHTGLV-UHFFFAOYSA-N
Synonym 1-phenylcyclopropanecarboxylic acid, 1-phenyl-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-phenyl, 1-phenyl-cyclopropanecarboxylic acid, 1-phenyl-1-cyclopropanecarboxylicacid, pubchem15554, acmc-1b2kn, phenylcyclopropanecarboxylic acid, 1-phenyl-cyclopropanecarboxlic acid, 1-phenylcyclo-propanecarboxylic acid
PubChem CID 80206
IUPAC Name 1-phenylcyclopropane-1-carboxylic acid
SMILES C1CC1(C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 85°C to 89°C
Quantity 5 g
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material 1-Phenylcyclopropanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.