missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylimidazole, 99%, Thermo Scientific™

Product Code. 11392866
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392866 1 g 1g
11302876 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392866 Supplier Thermo Scientific Alfa Aesar Supplier No. L10579.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-phenylimidazole is a ligand for cytochrome P-450 and used as an inhibitor of microsomal oxidation. 1-Phenylimidazole is a suitable reagent used to investigate its effect on the citrulline formation by bovine brain nitric-oxide synthase. 1-Phenylimidazole is reported to be inhibitor of calmodulin-dependent nitric-oxide synthase from bovine brain and GHs pituitary cells.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7164-98-9
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00041204
InChI Key SEULWJSKCVACTH-UHFFFAOYSA-N
Synonym 1-phenyl-1h-imidazole, phenylimidazole, n-phenylimidazole, 1h-imidazole, 1-phenyl, 1h-imidazole, phenyl, n-phenyl imidazole, pubchem12611, acmc-20aos8, 1-phenylimidazole, 1-phenyl-1h-imidazole #
PubChem CID 81595
SMILES C1=CN(C=N1)C1=CC=CC=C1

Specifications

Melting Point 13°C
Density 1.14
Boiling Point 142°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.6
Quantity 1 g
Beilstein 112356
Solubility Information Slightly soluble in water.
IUPAC Name 1-phenyl-1H-imidazole
Formula Weight 144.18
Percent Purity 99%
Chemical Name or Material 1-Phenylimidazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.