missing translation for 'onlineSavingsMsg'
Learn More

1-Propionylimidazole, TRC

Product Code. 30427390
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427390 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30427390 Supplier Toronto Research Chemicals Supplier No. TRCP839508250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Propionylimidazole
CAS 4122-52-5
Molecular Formula C6H8N2O
Quantity 250 mg
Synonym 1-(1-Oxopropyl)-1H-imidazole, 1-(1H-Imidazol-1-yl)-1-propanone, 1-Propionylimidazole, N-Propionylimidazole, Propionylimidazolide
InChI Formula InChI=1S/C6H8N2O/c1-2-6(9)8-4-3-7-5-8/h3-5H,2H2,1H3
SMILES CCC(=O)n1ccnc1
IUPAC Name 1-imidazol-1-ylpropan-1-one
Molecular Weight (g/mol) 124.14
Formula Weight 124.0637
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.