missing translation for 'onlineSavingsMsg'
Learn More

1-Propylamine, TRC

Product Code. 30414143
Change view
Click to view available options
Quantity:
50 mL
250 mL
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30414143 50 mL 50mL
30409294 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30414143 Supplier Toronto Research Chemicals Supplier No. TRCP83370050ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material n-Propylamine
CAS 107-10-8
Molecular Formula C3 H9 N
Quantity 50 mL
Synonym 1-Propanamine, Propylamine (8CI), (n-Propan-1-yl)amine, 1-Aminopropane, 1-Propylamine, 3-Aminopropane, Mono-n-propylamine, Monopropylamine, NSC 7490, Propan-1-amine, Propan-1-ylamine, n-Propylamine
InChI Formula InChI=1S/C3H9N/c1-2-3-4/h2-4H2,1H3
SMILES CCCN
IUPAC Name propan-1-amine
Molecular Weight (g/mol) 59.11
Formula Weight 59.0735
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.