Learn More
1-Tosyl-1H-pyrrole-2-carbaldehyde, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 102619-05-6 |
| Molecular Formula | C12H11NO3S |
| MDL Number | MFCD00671580 |
| Quantity | 5 g |
| SMILES | O=CC1=CC=CN1S(=O)(C2=CC=C(C)C=C2)=O |
| Molecular Weight (g/mol) | 249.29 |
| Percent Purity | 98% |
| Chemical Name or Material | 1-Tosyl-1H-pyrrole-2-carbaldehyde |
Product Title
Vous avez repéré une opportunité d'amélioration ?