missing translation for 'onlineSavingsMsg'
Learn More

1-Vinyl Ramipril, TRC

Product Code. 30455222
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
2.5 mg
Unit Size:
0.5mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455222 0.5 mg 0.5mg
30456860 1 mg 1mg
30457028 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455222 Supplier Toronto Research Chemicals Supplier No. TRCV1110450.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Vinyl Ramipril
Analyte or Component Names 1-Vinyl Ramipril
Shipping Condition Room Temperature
Molecular Formula C24 H32 N2 O5
Synonym (2S,3aS,6aS)-1-((S)-2-(((S)-1-(Allyloxy)-1-oxo-4-phenylbutan-2-yl)amino)propanoyl)octahydrocyclopenta[b]pyrrole-2-carboxylic Acid
InChI Formula InChI=1S/C24H32N2O5/c1-3-14-31-24(30)19(13-12-17-8-5-4-6-9-17)25-16(2)22(27)26-20-11-7-10-18(20)15-21(26)23(28)29/h3-6,8-9,16,18-21,25H,1,7,10-15H2,2H3,(H,28,29)/t16-,18-,19-,20-,21-/m0/s1
Solution Type Neat
SMILES C[C@H](N[C@@H](CCc1ccccc1)C(=O)OCC=C)C(=O)N2[C@H]3CCC[C@H]3C[C@H]2C(=O)O
IUPAC Name (2S,3aS,6aS)-1-[(2S)-2-[[(2S)-1-oxo-4-phenyl-1-prop-2-enoxybutan-2-yl]amino]propanoyl]-3,3a,4,5,6,6a-hexahydro-2H-cyclopenta[b]pyrrole-2-carboxylic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 428.521
Quantity 0.5 mg
Formula Weight 428.231
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.