Learn More
10,11-Dihydro-10-hydroxy Carbamazepine-D4 (Major), TRC
Product Code. 30381682
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30381682 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30381682 Supplier Toronto Research Chemicals Supplier No. TRCD4491371MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 10,11-Dihydro-10-hydroxycarbazepine-D4 (Major)
CAS 1020719-39-4
Purity Grade Notes HPLC
Molecular Formula C15 2H4 H10 N2 O2
Quantity 1 mg
Synonym Licarbazepine D4
InChI Formula InChI=1S/C15H14N2O2/c16-15(19)17-12-7-3-1-5-10(12)9-14(18)11-6-2-4-8-13(11)17/h1-8,14,18H,9H2,(H2,16,19)/i1D,3D,5D,7D
SMILES [2H]c1c([2H])c([2H])c2N(C(=O)N)c3ccccc3C(O)Cc2c1[2H]
IUPAC Name 1,2,3,4-tetradeuterio-6-hydroxy-5,6-dihydrobenzo[b][1]benzazepine-11-carboxamide
Molecular Weight (g/mol) 258.31
Formula Weight 258.1306
Percent Purity >95