missing translation for 'onlineSavingsMsg'
Learn More

10-Methyl Docetaxel, TRC

Product Code. 30469711
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30469711 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30469711 Supplier Toronto Research Chemicals Supplier No. TRCM3200201MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 10-Methyl Docetaxel
CAS 160084-81-1
Molecular Formula C44 H55 N O14
Quantity 1 mg
Synonym 10-Methyl Docetaxel
InChI Formula InChI=1S/C44H55NO14/c1-23-27(56-38(51)32(48)31(25-16-12-10-13-17-25)45-39(52)59-40(3,4)5)21-44(53)36(57-37(50)26-18-14-11-15-19-26)34-42(8,35(49)33(54-9)30(23)41(44,6)7)28(47)20-29-43(34,22-55-29)58-24(2)46/h10-19,27-29,31-34,36,47-48,53H,20-22H2,1-9H3,(H
SMILES CO[C@H]1C(=O)[C@]2(C)[C@@H](O)C[C@H]3OC[C@@]3(OC(=O)C)[C@H]2[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](OC(=O)[C@H](O)[C@@H](NC(=O)OC(C)(C)C)c6ccccc6)C(=C1C5(C)C)C
Molecular Weight (g/mol) 821.906
Formula Weight 821.362
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.