Learn More
Fisherbrand™ 100-Well Microtube Storage Boxes
Product Code. 15589811 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Color:
Blue
Green
Yellow
Unit Size:
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15589811 Yellow Pack of 5
15569811 Blue Pack of 5
15579811 Green Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15589811
  • ONLINE EXCLUSIVE PRICE
    73.25€
    30.19€ / Pack of 5
    Save 43.06€
    58% Off
View all promotions
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Stores small vials and microtubes. Fisherbrand™ 100-Well Microtube Storage Boxes keep contents in place with snap-close hinged lid.
  • For larger storage requirements of 1.5/2.0mL microtubes or cryogenic vials.
  • Easy-open hinged lid with snap clasp
  • 100 wells
  • Well dia.: 12.3mm
  • Dimensions: 141 x 151 x 57mm
  • Autoclavable
Specifications
Closure Type Hinged Lid
Color Yellow
Material Polypropylene
Shape Rectangular
Type Cryo Storage Box
For Use With (Application) Microtube Storage
Holds 100 (1.5 to 2 mL Tubes)
Includes Snap-Close Hinged Lid
Quantity 5/Pk.
Tube Array 10 x 10
Dimensions (L x W x H) 141 x 151 x 57 mm
Length (Metric) 141 mm
Width (Metric) 151 mm
Height (Metric) 57 mm
Diameter (Metric) Well 12.3 mm
Weight (Metric) 0.93 kg