Learn More
1000 NTU Ratio Turbidity Standard, Reagecon
BIOPHARMA
Product Code. 12736935 Shop All Reagecon Products
Change view
Click to view available options
Volume (Metric):
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12736935 500 mL 500mL
11536593 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12736935 Supplier Reagecon Supplier No. CRSR1000500
  • ONLINE EXCLUSIVE PRICE
    555.00€
    491.00€ / 500mL
    Save 64.00€
    11% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Suspension of extremely accurate (± 1%) polymer microspheres with guaranteed shelf life stability even after opening the bottle.
  • Turbidity Reference Standard, Turbidity Reference Material.
  • Designed and formulated to overcome the adverse safety, accuracy and stability issues experienced with other turbidity standards, primarily formazin-based standards.
  • Certified
  • NIST traceable
  • Non-toxic, non-carcinogenic
  • Highly accurate
  • Stable
  • EPA approved
Specifications
For Use With (Equipment) Ratio Instruments
Description Liquid format in an HDPE Bottle GL45
Accuracy Accuracy ± 1% NTU
Turbidity 1000 NTU ± 1%
Volume (Metric) 500 mL