missing translation for 'onlineSavingsMsg'
Learn More

(3-(tert-Butyl)-1H-pyrazol-5-yl)methanol, TRC

Product Code. 30180413
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180413 100 mg 100mg
30179754 500 mg 500mg
30180124 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180413 Supplier Toronto Research Chemicals Supplier No. TRCT221696100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 493038-53-2
Molecular Formula C8H14N2O
Molecular Weight (g/mol) 154.21
Synonym (5-tert-Butyl-1H-pyrazol-3-yl)-methanol
IUPAC Name (5-tert-butyl-1H-pyrazol-3-yl)methanol
SMILES CC(C)(C)c1cc(CO)n[nH]1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H14N2O/c1-8(2,3)7-4-6(5-11)9-10-7/h4,11H,5H2,1-3H3,(H,9,10)
Formula Weight 154.1106
Chemical Name or Material (3-(tert-Butyl)-1H-pyrazol-5-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.