missing translation for 'onlineSavingsMsg'
Learn More

Ketoprofen 2,3-Butylene Glycol Ester, Mikromol™

Product Code. 18022280
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18022280 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18022280 Supplier Mikromol Supplier No. MM0001.35

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ketoprofen 2,3-Butylene Glycol Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ketoprofen 2,3-Butylene Glycol Ester
CAS 94561-67-8
Analyte or Component Names Ketoprofen 2,3-Butylene Glycol Ester
Shipping Condition Room Temperature
Molecular Formula C20 H22 O4
Certifications/Compliance ISO 9001
Synonym Ketoprofen 2,3-Butylene Glycol Ester, 3-Benzoyl-alpha-methylbenzeneacetic acid 2-hydroxy-1-methylpropyl ester
InChI Formula InChI=1S/C20H22O4/c1-13(20(23)24-15(3)14(2)21)17-10-7-11-18(12-17)19(22)16-8-5-4-6-9-16/h4-15,21H,1-3H3
SMILES CC(O)C(C)OC(=O)C(C)c1cccc(c1)C(=O)c2ccccc2
IUPAC Name 3-hydroxybutan-2-yl 2-(3-benzoylphenyl)propanoate
Molecular Weight (g/mol) 326.39
Quantity 100 mg
Formula Weight 326.1518 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.