missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl 3-Methoxy-4-nitrobenzoate, TRC

Product Code. 30180028
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180028 100 mg 100mg
30179826 500 mg 500mg
30179925 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180028 Supplier Toronto Research Chemicals Supplier No. TRCS692043100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2006277-78-5
Molecular Formula C12H10N2O7
Molecular Weight (g/mol) 294.22
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 3-methoxy-4-nitrobenzoate
SMILES COc1cc(ccc1[N+](=O)[O-])C(=O)ON2C(=O)CCC2=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H10N2O7/c1-20-9-6-7(2-3-8(9)14(18)19)12(17)21-13-10(15)4-5-11(13)16/h2-3,6H,4-5H2,1H3
Formula Weight 294.0488
Chemical Name or Material N-Succinimidyl 3-Methoxy-4-nitrobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.