missing translation for 'onlineSavingsMsg'
Learn More

DLL3 Antibody (DLL3/9361), Novus Biologicals Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-45051

Product Code. 30448890

  • 593.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DLL3 Monoclonal antibody specifically detects DLL3 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DLL3
Monoclonal
1 mg/mL
Immunohistochemistry : 1-2ug/mL
delta (Drosophila)-like 3, Delta3, delta-like 3 (Drosophila), delta-like protein 3, Drosophila Delta homolog 3, SCDO1delta3
Recombinant fragment (around aa400-618) of human DLL3 protein (Uniprot: Q9NYJ7). Exact sequence is proprietary.
100 μg
Cell Biology, Cell Cycle and Replication, Neuroscience
10683
Store at -20 to -84°C. Avoid freeze-thaw cycles.
Purified
Immunohistochemistry
DLL3/9361
Unconjugated
10 mM PBS
Mouse
Protein A or G purified
RUO
Primary
Human
Antibody
IgG2c κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.