missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRAS Antibody (OTI2B7), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44764
This item is not returnable.
View return policy
Description
RRAS Monoclonal antibody specifically detects RRAS in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| RRAS | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized powder ( 1X PBS, pH 7.3,) | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| OTI2B7 | |
| Western Blot 1:500-2000 | |
| Oncogene RRAS, p23, ras-related protein R-Ras, related RAS viral (r-ras) oncogene homolog | |
| Full length human recombiant protein of human RRAS (NP_006261) produced in HEK293T cell. | |
| 100 μg | |
| Signal Transduction | |
| 6237 | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction