missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBS2 Antibody (9F10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42932
This item is not returnable.
View return policy
Description
BBS2 Monoclonal antibody specifically detects BBS2 in Canine,Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| BBS2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Canine, Human | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 9F10 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/ Immunofluorescence 1:20 - 1:200 | |
| Bardet-Biedl syndrome 2, Bardet-Biedl syndrome 2 protein, BBS, MGC20703 | |
| Bardet-Biedl syndrome 2 - (Uniprot# Q9BXC9) | |
| 100 μg | |
| Vision | |
| 583 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction