missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calponin 2 Antibody (4C9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42944
This item is not returnable.
View return policy
Description
Calponin 2 Monoclonal antibody specifically detects Calponin 2 in Human,Porcine samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| Calponin 2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Pig | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| 4C9 | |
| Western Blot 1:1000 - 1:4000, ELISA, Immunohistochemistry 1:500 - 1:4000 | |
| calponin 2, Calponin H2, smooth muscle, calponin-2, Neutral calponin | |
| calponin 2 - (Uniprot# Q99439) | |
| 100 μg | |
| Signal Transduction | |
| 1265 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction