missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Profilin 2 Antibody (5D4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43015
This item is not returnable.
View return policy
Description
Profilin 2 Monoclonal antibody specifically detects Profilin 2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| Profilin 2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| 5D4 | |
| Western Blot 1:1000 - 1:5000, ELISA, Immunohistochemistry 1:50 - 1:500 | |
| D3S1319E, PFL, profilin 2, Profilin II, profilin-2 | |
| profilin 2 - (Uniprot# P35080) | |
| 100 μg | |
| Cell Biology, Cellular Markers, Signal Transduction, Stem Cell Markers | |
| 5217 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction