missing translation for 'onlineSavingsMsg'
Learn More

RPL3 Antibody (3F3), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-43030

Product Code. 30279526

  • 473.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

RPL3 Monoclonal antibody specifically detects RPL3 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RPL3
Monoclonal
Unconjugated
PBS, pH 7.3, 50% glycerol
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
3F3
Western Blot 1:1000 - 1:8000, ELISA, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/ Immunofluorescence 1:20 - 1:100
60S ribosomal protein L3, HIV-1 TAR RNA-binding protein B, MGC104284, ribosomal protein L3, TARBP-B
ribosomal protein L3 - (Uniprot# P39023)
100 μg
Immunology
6122
Store at -20°C. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.