missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transketolase Antibody (4E2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43051
This item is not returnable.
View return policy
Description
Transketolase Monoclonal antibody specifically detects Transketolase in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Transketolase | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 400 | |
| Western Blot 1:500 - 1:5000, ELISA, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/ Immunofluorescence 1:20 - 1:200 | |
| EC 2.2.1.1, FLJ34765, TK, TKT1, transketolase | |
| transketolase - (Uniprot# P29401) | |
| 100 μg | |
| Stem Cell Markers | |
| 7086 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction