missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ EIF2AK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29375
This item is not returnable.
View return policy
Description
EIF2AK1 Polyclonal antibody specifically detects EIF2AK1 in Human samples. It is validated for Immunoprecipitation
Specifications
| EIF2AK1 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| EC 2.7.11, eukaryotic translation initiation factor 2-alpha kinase 1, HCR, heme sensitive initiation factor 2a kinase, Heme-controlled repressor, Heme-regulated eukaryotic initiation factor eIF-2-alpha kinase, Heme-regulated inhibitor, heme-regulated initiation factor 2-alpha kinase, heme-regulated repressor, Hemin-sensitive initiation factor 2-alpha kinase, HRIEC 2.7.11.1, KIAA1369heme regulated initiation factor 2 alpha kinase | |
| between 525 and 575 | |
| 100 μg | |
| Protein Kinase | |
| 27102 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction