Learn More
Novus Biologicals™ PRMT10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29238
This item is not returnable.
View return policy
Description
PRMT10 Polyclonal antibody specifically detects PRMT10 in Human samples. It is validated for Immunoprecipitation
Specifications
| PRMT10 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| EC 2.1.1.-, FLJ46629, protein arginine methyltransferase 10 (putative), putative protein arginine N-methyltransferase 10 | |
| Between 1 and 50 | |
| 100 μg | |
| Cell Biology | |
| 90826 | |
| Store at 4°C. Do not freeze. | |
| IgG |