missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Influenza A H3N2 M1 Polyclonal Antibody, FITC

Goat Polyclonal Antibody
Brand: Invitrogen™ PA173050
This item is not returnable.
View return policy
Description
The PA1-73050 antibody reacts with Influenza A matrix protein. PA1-73050 has been successfully used in Immunofluorescence applications. The PA1-73050 antibody was raised against Influenza A, Phillipines (H3N2).
Influenza A virus is an enveloped virus encoding 10 polypeptides. RNA segment 7 encodes for two proteins: M1 (matrix 1) and M2 (matrix 2). M1 protein is encoded by an mRNA that is colinear, while M2 protein is synthesized from spliced mRNA. M2 protein is a transmembrane protein composed of three Domains: 1) 24 residues representing the N-terminal region, 2) 19 hydro-phobic residues that serve as a membrane anchor, and 3) 54 residues near the C-terminal in the cytoplasmic domain. The M2 protein has been found to play a role in Influenza replication and assembly of virion particles. Further experimentation has demonstrated that this protein is an acid-activated ion channel for virus replication.
Specifications
| Influenza A H3N2 M1 | |
| Polyclonal | |
| FITC | |
| H3N2; H3N2 M1; H3N2 Matrix protein; Influenza H3N2 | |
| Influenza A, Phillipines (H3N2). | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
| Immunocytochemistry | |
| 4-5 mg/mL | |
| PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 | |
| Goat | |
| IgG fraction | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction