missing translation for 'onlineSavingsMsg'
Learn More

10H-Phenothiazine 5-Oxide, TRC

Product Code. 30420055
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420055 500 mg -
30421033 50 mg -
30412584 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30420055 Supplier Toronto Research Chemicals Supplier No. TRCP318098500MG
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1207-71-2
Molecular Formula C12H9NOS
Molecular Weight (g/mol) 215.27
Synonym Phenothiazine 5-Oxide, NSC 3554, Phenothiazine S-Oxide
IUPAC Name 10H-phenothiazine 5-oxide
SMILES O=S1c2ccccc2Nc3ccccc13

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C12H9NOS/c14-15-11-7-3-1-5-9(11)13-10-6-2-4-8-12(10)15/h1-8,13H
Formula Weight 215.0405
Chemical Name or Material 10H-Phenothiazine 5-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.