missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Verapamil-d6 Hydrochloride, (R)-(+)-Verapamil-d6 Hydrochloride, TRC

Product Code. 30240722
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240722 10 mg 10mg
30240502 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30240722 Supplier Toronto Research Chemicals Supplier No. TRCV12499210MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C27H32D6N2O4 • HCl
Molecular Weight (g/mol) 460.653646
Synonym (αR)-α-[3-[[2-(3,4-Dimethoxyphenyl)ethyl]methylamino]propyl]-3,4-dimethoxy-α-(1-methylethyl)benzeneacetonitrile-d6 Hydrochloride, (R)-Verapamil-d6 Hydrochloride, Dexverapamil-d6 Hydrochloride, LU 33925-d6, NSC 632821-d6;
SMILES COC1=C(OC)C=CC(CCN(C)CCC[C@](C(C([2H])([2H])[2H])C([2H])([2H])[2H])(C#N)C2=CC=C(OC)C(OC)=C2)=C1.Cl

Specifications

CAS 1795786-09-2
Quantity 10 mg
InChI Formula InChI=1S/C27H38N2O4.ClH/c1-20(2)27(19-28,22-10-12-24(31-5)26(18-22)33-7)14-8-15-29(3)16-13-21-9-11-23(30-4)25(17-21)32-6;/h9-12,17-18,20H,8,13-16H2,1-7H3;1H/t27-;/m1./s1/i1D3,2D3;
Formula Weight 496.2975
Chemical Name or Material (R)-(+)-Verapamil-d6 Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.