missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tildipirosin-d10 (90%), TRC
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Specifications
Specifications
| Chemical Name or Material | Tildipirosin-d10 |
| Molecular Formula | C41H61D10N3O8 |
| Quantity | 10 mg |
| Synonym | 20-Deoxo-23-deoxy-5-O-[3,6-dideoxy-3-(dimethylamino)-β-D-glucopyranosyl]-20,23-di-1-piperidinyltylonolide-d10 |
| InChI Formula | InChI=1S/C41H71N3O8/c1-8-35-32(26-44-20-13-10-14-21-44)23-27(2)15-16-33(45)28(3)24-31(17-22-43-18-11-9-12-19-43)40(29(4)34(46)25-36(47)51-35)52-41-39(49)37(42(6)7)38(48)30(5)50-41/h15-16,23,28-32,34-35,37-41,46,48-49H,8-14,17-22,24-26H2,1-7H3/b16-15+,27-23+/t28-,29+,30-,31+,32-,34-,35-,37+,38-,39?,40-,41+/m1/s1/i10D2,13D2,14D2,20D2,21D2 |
| SMILES | O=C(C[C@@H](O)[C@H](C)[C@@H](O[C@H]1C(O)[C@@H](N(C)C)[C@H](O)[C@@H](C)O1)[C@@H](CCN2CCCCC2)C[C@H]3C)O[C@H](CC)[C@@H](CN4C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C4([2H])[2H])/C=C(C)/C=C/C3=O |
| IUPAC Name | (4R,5S,6S,7R,9R,11E,13E,15R,16R)-6-(((2R,4S,5S,6R)-4-(dimethylamino)-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-16-ethyl-4-hydroxy-5,9,13-trimethyl-7-(2-(piperidin-1-yl)ethyl)-15-((piperidin-1-yl-d10)methyl)oxacyclohexadeca-11,13-diene-2,10-dione |
| Molecular Weight (g/mol) | 744.08 |
| Formula Weight | 743.59 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction