missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Varenicline-d2,15N2 Dihydrochloride, TRC
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Specifications
Specifications
| Chemical Name or Material | Varenicline-d2,15N2 Dihydrochloride |
| Molecular Formula | C13H13D2Cl2N15N2 |
| Quantity | 10 mg |
| Synonym | 7,8,9,10-Tetrahydro-6,10-methano-6H-pyrazino[2,3-h][3]benzazepine-d2,15N2 Dihydrochloride |
| InChI Formula | InChI=1S/C13H13N3.2ClH/c1-2-16-13-5-11-9-3-8(6-14-7-9)10(11)4-12(13)15-1;;/h1-2,4-5,8-9,14H,3,6-7H2;2*1H/i1D,2D,15+1,16+1;; |
| SMILES | [2H]C1=[15N]C(C=C(C2CNCC3C2)C3=C4)=C4[15N]=C1[2H].Cl.Cl |
| Molecular Weight (g/mol) | 288.18 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction