Learn More
Thermo Scientific™ T4 DNA Ligase Buffer (10X)
Product Code. 11889622 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1.5 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11889622 1.5 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11889622 Supplier Thermo Scientific™ Supplier No. B69
  • ONLINE EXCLUSIVE PRICE
    22.90€
    22.65€ / 1.5mL
    Save 0.25€
    1% Off
View all promotions
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Safely ligate samples with this buffer, for use with T4 DNA Ligase and tested for the absence of endo- and exodeoxyribonucleases and ribonucleases.
10X T4 DNA Ligase buffer is used with T4 DNA Ligase.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 10X
Quantity 1.5 mL
Product Type T4 DNA Ligase Buffer