missing translation for 'onlineSavingsMsg'
Learn More

11-Hydroxylauric Acid, TRC

Product Code. 30180236
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180236 50 mg 50mg
30179989 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180236 Supplier Toronto Research Chemicals Supplier No. TRCH94373550MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32459-66-8
Molecular Formula C12 H24 O3
Molecular Weight (g/mol) 216.32
Synonym 11-Hydroxydodecanoic Acid
IUPAC Name 11-hydroxydodecanoic acid
SMILES CC(O)CCCCCCCCCC(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H24O3/c1-11(13)9-7-5-3-2-4-6-8-10-12(14)15/h11,13H,2-10H2,1H3,(H,14,15)
Formula Weight 216.1725
Chemical Name or Material 11-Hydroxylauric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.