missing translation for 'onlineSavingsMsg'
Learn More

11-Keto Oxcarbazepine, TRC

Product Code. 30007406
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007406 5 mg 5mg
30011007 10 mg 10mg
30006608 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007406 Supplier Toronto Research Chemicals Supplier No. TRCK1975005MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 537693-29-1
Molecular Formula C15 H10 N2 O3
Molecular Weight (g/mol) 266.25
Synonym 10,11-Dihydro-10,11-dioxo-5H-dibenz[b,f]azepine-5-carboxamide, Oxcarbazepine Impurity 9
IUPAC Name 5,6-dioxobenzo[b][1]benzazepine-11-carboxamide
SMILES NC(=O)N1c2ccccc2C(=O)C(=O)c3ccccc13

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C15H10N2O3/c16-15(20)17-11-7-3-1-5-9(11)13(18)14(19)10-6-2-4-8-12(10)17/h1-8H,(H2,16,20)
Formula Weight 266.0691
Chemical Name or Material 11-Keto Oxcarbazepine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.