missing translation for 'onlineSavingsMsg'
Learn More

Iodine, crystalline, 99.5%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 014248.14

Product Code. 11307827

  • 51.00€ / 25g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

7553-56-2
99.5%
MFCD00011355,MFCD00164163
PNDPGZBMCMUPRI-UHFFFAOYSA-N
diiodine
807
253.81
Iodine, crystalline
25g
I2
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
II
253.81
CHEBI:17606
4.93 g/mL
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.