Learn More
Labconco™ Drying Chamber for FreeZone™ Freeze-Dry Systems, PTFE-coated
Product Code. 15968132 Shop All Labconco Corporation Products
Change view
Click to view available options
No. of Ports:
12
16
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Ports Unit Size
15968132 16 Each
15954650 12 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15968132 Supplier Labconco™ Supplier No. 7522901
  • ONLINE EXCLUSIVE PRICE
    4325.00€
    3523.22€ / Pack of 1
    Save 801.78€
    18% Off
View all promotions
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket
Aggiungi al carrello
missing translation for 'validMainframeMsg'
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Drying Chamber with corrosion-resistant interior for use on FreeZone™ Freeze Dryers.
  • Drying chamber made of type 304 stainless steel with corrosion-resistant PTFE-coated interior
  • Includes acrylic lid with neoprene gasket, latching pin, and neoprene valves with molded plastic knobs for 1/2 in. and 3/4 in. adapters
  • For use with FreeZone™ 1, 2.5, 4.5, 6, 8, 12, or 18 Liter Freeze Dryers
Specifica
Type Drying Chamber
No. of Shelves 0
Material PTFE-coated Stainless Steel
No. of Ports 16
For Use With (Application) To provide a means to attach flasks to freeze dry system or contain samples in bulk
For Use With (Equipment) FreeZone™ Freeze Dryers
Diameter (English) 13 in.
Height (English) 13 in.
Shipping Weight (English) 21 lb.
Weight (English) 20 lb.
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.