missing translation for 'onlineSavingsMsg'
Learn More

13-Ethyl-17-hydroxy-11-methylidene-18,19-dinor-17alpha-pregn-4-en-20-yn-3-one (3-Ketodesogestrel), Mikromol™

Product Code. 30050049
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050049 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050049 Supplier Mikromol Supplier No. MM1024.030025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

13-Ethyl-17-hydroxy-11-methylidene-18,19-dinor-17alpha-pregn-4-en-20-yn-3-one (3-Ketodesogestrel)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Ketodesogestrel
CAS 54048-10-1
Analyte or Component Names 3-Ketodesogestrel
Shipping Condition Room Temperature
Molecular Formula C22 H28 O2
Certifications/Compliance ISO 17034
Synonym Etonogestrel, 3-Keto-desogestrel, Desogestrel Imp. D (EP), Desogestrel USP Related Compound C, Desogestrel USP RC C, Desogestrel Impurity D, 13-Ethyl-17-hydroxy-11-methylidene-18,19-dinor-17alpha-pregn-4-en-20-yn-3-one, 18,19-Dinorpregn-4-en-20-yn-3-one, 13-ethyl-17-hydroxy-11-methylene-, (17alpha)-, (17alpha)-13-Ethyl-17-hydroxy-11-methylene-18,19-dinorpregn-4-en-20-yn-3-one, 17-Ethynyl-17beta-hydroxy-18-methyl-11-methyleneestr-4-en-3-one, 3-Keto-Org 2969, 3-Ketodesogestrel, 3-Oxodesogestrel, Implanon, Nexplanon, Org 3236, Desogestrel Related Compound C, Desogestrel Impurity D
InChI Formula InChI=1S/C22H28O2/c1-4-21-13-14(3)20-17-9-7-16(23)12-15(17)6-8-18(20)19(21)10-11-22(21,24)5-2/h2,12,17-20,24H,3-4,6-11,13H2,1H3/t17-,18-,19-,20+,21-,22-/m0/s1
SMILES CC[C@]12CC(=C)[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@]2(O)C#C
IUPAC Name (8S,9S,10R,13S,14S,17R)-13-ethyl-17-ethynyl-17-hydroxy-11-methylidene-2,6,7,8,9,10,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 324.46
Quantity 25 mg
Formula Weight 324.2089 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.