missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV16 E1+E4 Antibody (HPV16 E1/E4), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08319PE
This item is not returnable.
View return policy
Description
HPV16 E1+E4 Monoclonal antibody specifically detects HPV16 E1+E4 in Virus samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| HPV16 E1+E4 | |
| Monoclonal | |
| PE | |
| ATP dependent helicase E1, E1, E4, HPV 16 ATP dependent helicase E1, HPV 16 E1, HPV 16 E4, HPV 16 replication protein E1, HPV16 ATP dependent helicase E1, HPV16 E1, HPV16 E4, HPV16 replication protein E1, Human papilloma virus type 16, Human papilloma virus type 16 ATP dependent helicase E1, Human papilloma virus type 16 E1, Human papilloma virus type 16 E4, Human papilloma virus type 16 replication protein E1, Human papillomavirus type 16, Human papillomavirus type 16 ATP dependent helicase E1, Human papillomavirus type 16 E1, Human papillomavirus type 16 E4, Human papillomavirus type 16 replication protein E1, Replication protein, Replication protein E1 | |
| Recombinant human HPV16 E1+E4 fragment (aa 36-41). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| HPV16 E1/E4 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction