missing translation for 'onlineSavingsMsg'
Learn More

14-3-3 zeta Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

213.00€ - 351.00€

Specifications

Antigen 14-3-3 zeta
Concentration 0.5 mg/mL
Dilution Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18169981
View Documents
Novus Biologicals
NBP2-24593SS
0.025 mg
213.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18160342
View Documents
Novus Biologicals
NBP2-24593
0.1 mg
351.00€
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

14-3-3 zeta Polyclonal specifically detects 14-3-3 zeta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

14-3-3 zeta
Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Polyclonal
Purified
RUO
Human, Mouse, Rat, Bovine, Canine, Primate
KCIP-1MGC126532,14-3-3-zeta, MGC111427, MGC138156, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, deltapolypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zetapolypeptide, zeta polypeptide
YWHAZ
IgG
Protein A purified
The amino acid sequence used as immunogen for the antibody is 100% homologous in human and 94% homologous in mouse, rat and bovine.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cell Cycle and Replication
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
7534
A portion of amino acids 200-245 of human 14-3-3 zeta was used as the immunogen for the antibody.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.