missing translation for 'onlineSavingsMsg'
Learn More

15α-Acetoxy-gestodene, TRC

Product Code. 30327161
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327161 5 mg 5mg
30328688 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30327161 Supplier Toronto Research Chemicals Supplier No. TRCA1655805MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 13-Ethyl-17-hydroxy-3-oxo-18,19-dinor-17Alpha-pregn-4-en-20-yn-15Alpha-yl Acetate
CAS 267650-77-1
Molecular Formula C23 H30 O4
Quantity 5 mg
Synonym 18,19-Dinorpregn-4-en-20-yn-3-one, 15-(acetyloxy)-13-ethyl-17-hydroxy-, (15α,17α)- (9CI), 13-Ethyl-17-hydroxy-3-oxo-18,19-dinor-17α-pregn-4-en-20-yn-15α-yl acetate, 15α-Acetoxy-gestodene, Gestodene Impurity F (EP)
InChI Formula InChI=1S/C23H30O4/c1-4-22-11-10-18-17-9-7-16(25)12-15(17)6-8-19(18)21(22)20(27-14(3)24)13-23(22,26)5-2/h2,12,17-21,26H,4,6-11,13H2,1,3H3/t17-,18+,19+,20-,21+,22-,23-/m0/s1
SMILES CC[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1[C@H](C[C@@]2(O)C#C)OC(=O)C
IUPAC Name [(8R,9S,10R,13S,14S,15S,17R)-13-ethyl-17-ethynyl-17-hydroxy-3-oxo-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-15-yl] acetate
Molecular Weight (g/mol) 370.482
Formula Weight 370.214
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.