missing translation for 'onlineSavingsMsg'
Learn More

15-Methylhexadeca Sphinganine, TRC

Product Code. 30003831
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003831 5 mg 5mg
30004630 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30003831 Supplier Toronto Research Chemicals Supplier No. TRCM3119705MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26048-10-2
Molecular Formula C17 H37 N O2
Molecular Weight (g/mol) 287.48
Synonym [R-(R*,S*)]-2-Amino-15-methyl-1,3-hexadecanediol, D-erythro- 2-Amino-15-methyl-1,3-hexadecanediol, D-erythro-C17-iso-Sphinganine
IUPAC Name (2S,3R)-2-amino-15-methyl-hexadecane-1,3-diol
SMILES CC(C)CCCCCCCCCCC[C@@H](O)[C@@H](N)CO

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C17H37NO2/c1-15(2)12-10-8-6-4-3-5-7-9-11-13-17(20)16(18)14-19/h15-17,19-20H,3-14,18H2,1-2H3/t16-,17+/m0/s1
Formula Weight 287.2824
Chemical Name or Material 15-Methylhexadeca Sphinganine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.