missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK32A Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67528-1-IG-150UL
This item is not returnable.
View return policy
Description
The STK32A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human STK32A. This antibody recognizes human antigen. The STK32A antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| STK32A | |
| Monoclonal | |
| 1700 μg/mL | |
| MGC22688; YANK1 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human | |
| 50 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 1H2A6 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 202374 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction