Learn More
Invitrogen™ 158P1D7 Biosimilar Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558975
 View more versions of this product
Product Code. 30284152
  • 477.00€ / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term Agensys patent anti-158P1D7.
The gene 158P1D7, also known as the gene encoding the pentadecapeptide BPC 157, is derived from a partial sequence of the body protection compound (BPC) found in human gastric juice. BPC 157 is composed of 15 amino acids and has been identified for its significant therapeutic effects on wound healing and tissue regeneration. It is particularly noted for its role in accelerating the healing of various types of wounds, including tendons and ligaments, due to its ability to promote angiogenesis and modulate the inflammatory response. Its mechanisms involve interactions with growth factors and cellular signaling pathways, making it a promising candidate for treating musculoskeletal injuries and disorders.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
158P1D7 Biosimilar
Recombinant Monoclonal
Unconjugated
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Protein A
RUO
Human
Antibody
IgG2SA