Learn More
Brady™ Honeywell Voyager 1470G 2D Handheld Scanner
Product Code. 16844163 Shop All Brady Products
Change view
Click to view available options
Connection Type:
USB
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connection Type Unit Size
16844163 USB Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 16844163 Supplier Brady™ Supplier No. 306804
  • ONLINE EXCLUSIVE PRICE
    335.00€
    301.88€ / Pack of 1
    Save 33.12€
    9% Off
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Optimized for fast scanning of linear bar codes, including poorly printed and damaged codes, the Voyager 1470g2D enables enterprises to minimize manual data entry, leading to increased productivity and reduction in errors.
  • Enables enterprises to minimize manual data entry, leading to increased productivity and reduction in errors
Specifications
Description Imager Sensor
Connection Type USB
Power Supply 400 mA (at 5 V DC)
Alternate Part Number VOYAGER 1470G2D